Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tsh
- Entrez Gene ID
-
35430
- Gene Name
- teashirt
- Gene Aliases
-
CG1374, Dmel\CG1374, Dmel_CG1374, T shirt, TSH, Tsh, ae, ae[l], l(2)04319, l(2)B4-2-12
- Chromosome Location
-
Chr.2L: 21828593 - 21837011 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003866
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078891.3 |
NP_523615.2 |
2 - 3 |
4539 |
68 |
GenBank mRNA |
BT030762.1 |
- |
2 - 3 |
4537 |
68 |
View Full Product Details