Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adf1
Entrez Gene ID
47082
Gene Name
Adh transcription factor 1
Gene Aliases
ADF1, Adf 1, Adf-1, CG15845, Dmel\CG15845, Dmel_CG15845, adf-1, adf1, l(2)01349, l(2)04065, l(2)k14805, nal
Chromosome Location
Chr.2R: 6662265 - 6668085 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284249
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273801.1 NP_001260730.1 2 - 3 360 61
NM_165479.2 NP_724481.1 2 - 3 360 61
NM_165480.2 NP_724482.1 1 - 2 487 61
NM_206028.2 NP_995750.1 2 - 3 367 61
GenBank mRNA AY069387.1 - 1 - 2 461 61
BT088961.1 - 2 - 3 368 61
M37787.1 - 1 - 2 474 61

View Full Product Details