Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlf
Entrez Gene ID
36750
Gene Name
Myelodysplasia/myeloid leukemia factor
Gene Aliases
BcDNA:SD18148, CG8295, Dmel\CG8295, Dmel_CG8295, MLF, dMLF, dmlf, mlf
Chromosome Location
Chr.2R: 15935893 - 15940073 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034051
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169693.1 NP_001163164.1 1 - 2 159 63
NM_001259426.2 NP_001246355.1 1 - 2 153 63
NM_079029.5 NP_523753.1 1 - 2 153 63
NM_166123.4 NP_725502.1 1 - 2 159 63
NM_206125.2 NP_995847.1 1 - 2 159 63
GenBank mRNA AB043986.1 - 1 - 2 114 63
AY037049.1 - 1 - 2 125 63
AY118583.1 - 1 - 2 138 63

View Full Product Details