Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mute
Entrez Gene ID
2768848
Gene Name
muscle wasted
Gene Aliases
CG15610, CG33455, CG33456, CG34415, Dmel\CG34415, Dmel_CG34415, Mute
Chromosome Location
Chr.2R: 16948296 - 16955266 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085444
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103882.3 NP_001097352.1 3 - 4 2005 82

View Full Product Details