Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ktub
Entrez Gene ID
37400
Gene Name
king tubby
Gene Aliases
87D3S, CG9398, D-tulp, Dmel\CG9398, Dmel_CG9398, Dtulp, ESTS:87D3S, King-Tubby, King-tubby, TULP, Tulp, dTULP, dTulp, king-tubby, tulp
Chromosome Location
Chr.2R: 21101928 - 21123704 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015721
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137705.3 NP_611549.2 1 - 2 690 76
NM_206189.2 NP_995911.1 1 - 2 690 76
GenBank mRNA BT003201.1 - 1 - 2 688 76

View Full Product Details