Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scf
Entrez Gene ID
38145
Gene Name
supercoiling factor
Gene Aliases
BEST:CK02682, CG9148, CK02682, DCB-45, Dmel\CG9148, Dmel_CG9148, SCF, anon-EST:Posey14
Chromosome Location
Chr.3L: 1298672 - 1300389 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025682
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058044.5 NP_477392.1 1 - 2 528 58
NM_058045.4 NP_477393.1 1 - 2 314 58
GenBank mRNA AY070689.1 - 1 - 2 581 58
BT024934.1 - 1 - 2 502 58

View Full Product Details