Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gr64e
Entrez Gene ID
3771916
Gene Name
Gustatory receptor 64e
Gene Aliases
CG14988, CG32258, CG33157, DmGr64e, Dmel64e, Dmel\CG32258, Dmel_CG32258, GR64e, Gr64, Gr64A2, gr64e
Chromosome Location
Chr.3L: 4032141 - 4035703 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0045476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031935.2 NP_001027106.1 2 - 3 158 142

View Full Product Details