Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NAAT1
- Entrez Gene ID
-
31457
- Gene Name
- Nutrient Amino Acid Transporter 1
- Gene Aliases
-
CG3252, DmNAT1, Dmel\CG3252, Dmel_CG3252, anon-WO0149848.3, dmNAT1
- Chromosome Location
-
Chr.X: 5405238 - 5410849 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029762
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_131991.3 |
NP_572219.1 |
2 - 3 |
550 |
77 |
GenBank mRNA |
BT099637.1 |
- |
2 - 3 |
546 |
77 |
View Full Product Details