Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sytalpha
Entrez Gene ID
35068
Gene Name
Synaptotagmin alpha
Gene Aliases
CG5559, Dmel\CG5559, Dmel_CG5559, SYT-alpha, SYT3, Syt alpha, Syt3, dsytalpha, sytalpha
Chromosome Location
Chr.2L: 17592260 - 17604387 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261089
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201903.1 NP_001188832.1 3 - 4 677 76
NM_001201904.1 NP_001188833.1 3 - 4 935 76
NM_001299090.1 NP_001286019.1 3 - 4 935 76
NM_136009.2 NP_609853.1 3 - 4 783 76
GenBank mRNA BT060450.1 - 3 - 4 677 76

View Full Product Details