Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rdo
Entrez Gene ID
35077
Gene Name
reduced ocelli
Gene Aliases
CG15151, CT35050, Dmel\CG15151, Dmel_CG15151, PFE, Rdo
Chromosome Location
Chr.2L: 18012380 - 18070246 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0243486
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299095.1 NP_001286024.1 14 - 15 2404 73
NM_136017.3 NP_609861.1 14 - 15 2356 73
GenBank mRNA BT004502.1 - 14 - 15 2329 73

View Full Product Details