Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RecQ4
- Entrez Gene ID
-
53438
- Gene Name
- RecQ4 helicase
- Gene Aliases
-
CG7487, DmRECQ4, Dmel\CG7487, Dmel_CG7487, RECQ4, RECQL4, cg7487, dRecQ4
- Chromosome Location
-
Chr.3L: 8170640 - 8175978 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040290
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144350.2 |
NP_652607.1 |
3 - 4 |
4139 |
72 |
GenBank mRNA |
BT150480.1 |
- |
3 - 4 |
4059 |
72 |
View Full Product Details