The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144185.2 | NP_001137657.1 | 6 - 7 | 1202 | 59 |
NM_001144186.2 | NP_001137658.1 | 6 - 7 | 1202 | 59 | |
NM_001144187.2 | NP_001137659.1 | 6 - 7 | 1202 | 59 | |
NM_001144188.2 | NP_001137660.1 | 6 - 7 | 1425 | 59 | |
NM_001144189.2 | NP_001137661.1 | 6 - 7 | 1425 | 59 | |
NM_001169676.1 | NP_001163147.1 | 3 - 4 | 1218 | 59 | |
NM_001169677.1 | NP_001163148.1 | 6 - 7 | 1202 | 59 | |
NM_001169678.1 | NP_001163149.1 | 6 - 7 | 1425 | 59 | |
NM_001169679.2 | NP_001163150.1 | 3 - 4 | 1218 | 59 | |
NM_001259378.1 | NP_001246307.1 | 3 - 4 | 1218 | 59 | |
NM_001259379.1 | NP_001246308.1 | 3 - 4 | 1218 | 59 | |
NM_001259380.1 | NP_001246309.1 | 7 - 8 | 1220 | 59 | |
NM_001259381.1 | NP_001246310.1 | 7 - 8 | 1262 | 59 | |
NM_001259383.1 | NP_001246312.1 | 3 - 4 | 1218 | 59 | |
NM_001259384.1 | NP_001246313.1 | 3 - 4 | 1218 | 59 | |
NM_079009.3 | NP_523733.2 | 6 - 7 | 1425 | 59 | |
NM_166015.2 | NP_725335.1 | 6 - 7 | 1202 | 59 | |
NM_166016.2 | NP_725336.1 | 6 - 7 | 1202 | 59 | |
NM_166017.2 | NP_725337.1 | 6 - 7 | 1425 | 59 | |
NM_166018.3 | NP_725338.1 | 3 - 4 | 1218 | 59 | |
NM_166019.3 | NP_725339.1 | 3 - 4 | 1218 | 59 |