Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rpn6
- Entrez Gene ID
-
36638
- Gene Name
- Regulatory particle non-ATPase 6
- Gene Aliases
-
BcDNA.LD18931, BcDNA:LD18931, CG10149, CG5289, Dmel\CG10149, Dmel_CG10149, Pros44.5, RPN6, S9, dPSMD11, l(2)k00103, p42B, rpn6
- Chromosome Location
-
Chr.2R: 14750166 - 14752910 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028689
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166065.3 |
NP_725412.2 |
1 - 2 |
118 |
75 |
GenBank mRNA |
BT089015.1 |
- |
1 - 2 |
98 |
75 |
View Full Product Details