Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir51b
- Entrez Gene ID
-
246442
- Gene Name
- Ionotropic receptor 51b
- Gene Aliases
-
CG30081, DmelIR51b, Dmel\CG30081, Dmel_CG30081, IR51b
- Chromosome Location
-
Chr.2R: 15207993 - 15209786 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050081
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166080.1 |
NP_725440.1 |
1 - 1 |
1737 |
76 |
View Full Product Details