Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpo1
Entrez Gene ID
47611
Gene Name
Glycerophosphate oxidase 1
Gene Aliases
1(2) K05713, CG 8256, CG18786, CG8256, Dmel\CG8256, Dmel_CG8256, GPO, GPO-1, Gpo, Gpo-1, alpha-GPO, alphaGPO, anon-WO0118547.114, l(2)k05713
Chromosome Location
Chr.2R: 15857752 - 15862755 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022160
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103862.2 NP_001097332.1 5 - 6 816 60
NM_137219.3 NP_611063.1 5 - 6 605 60
NM_166118.2 NP_725495.1 5 - 6 612 60
NM_166119.2 NP_725496.1 5 - 6 671 60
GenBank mRNA AY051430.1 - 5 - 6 637 60

View Full Product Details