Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mub
Entrez Gene ID
40436
Gene Name
mushroom-body expressed
Gene Aliases
CG7437, Dmel\CG7437, Dmel_CG7437, MRE17, MUB, Mub, anon-WO0172774.33, anon-WO0172774.35, anon-WO0172774.36, anon-WO0172774.37, l(3)04093, l(3)j6B10, l(3)j8B3
Chromosome Location
Chr.3L: 21844788 - 21932702 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262737
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043154.4 NP_001036619.1 5 - 6 1212 67
NM_001170022.3 NP_001163493.1 5 - 6 1200 67
NM_001259944.2 NP_001246873.1 6 - 7 1312 67
NM_001259945.2 NP_001246874.1 5 - 6 1200 67
NM_001259946.2 NP_001246875.1 5 - 6 1212 67
NM_001259947.1 NP_001246876.1 6 - 7 1324 67
NM_001275268.1 NP_001262197.1 6 - 7 1312 67
NM_001275269.1 NP_001262198.1 6 - 7 1271 67
NM_001316446.1 NP_001303375.1 5 - 6 1212 67
NM_079484.6 NP_524208.1 5 - 6 1212 67
NM_168946.4 NP_730700.2 6 - 7 1312 67
NM_206423.5 NP_996145.1 5 - 6 1212 67
GenBank mRNA AY060403.1 - 5 - 6 1160 67
BT030417.1 - 5 - 6 1136 67
X99340.1 - - 1967 67

View Full Product Details