Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Or69a
Entrez Gene ID
2768964
Gene Name
Odorant receptor 69a
Gene Aliases
69F.1, 69a, 69b, CG17902, CG32116, CG33264, DOR82, DOR83, DmOr69a, Dmel\CG33264, Dmel_CG33264, Or69A, Or69aA, Or69aB, Or69ab, Or69b, or69a
Chromosome Location
Chr.3L: 12964360 - 12967733 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041622
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206347.1 NP_996069.1 2 - 3 1134 74
NM_206348.1 NP_996070.1 2 - 3 1134 74

View Full Product Details