The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038746.3 | NP_001033835.2 | 4 - 5 | 1744 | 58 |
NM_001258645.1 | NP_001245574.1 | 5 - 6 | 1167 | 58 | |
NM_001258647.1 | NP_001245576.1 | 5 - 6 | 1264 | 58 | |
NM_001272397.1 | NP_001259326.1 | 4 - 5 | 1744 | 58 | |
NM_001298081.1 | NP_001285010.1 | 4 - 5 | 1744 | 58 | |
NM_001298082.1 | NP_001285011.1 | 4 - 5 | 1744 | 58 |