Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cac
Entrez Gene ID
32158
Gene Name
cacophony
Gene Aliases
13, CAC, CG1522, CG15928, CG43368, Ca-alpha1A, Ca[[V]]2.1, Cac, Cac1, DMCA1A, Dmca1A, Dmcalpha1A, Dmel\CG43368, Dmel_CG43368, EE171, H18, H47, L13, P18, alpha1A, cac1, dCac1, l(1)11Aa, l(1)13, l(1)L13, ldg, nbA, nonB, opm18, opm47
Chromosome Location
Chr.X: 11931280 - 11985087 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263111
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014733.5 NP_001014733.1 9 - 10 1640 92
NM_001014734.4 NP_001014734.1 9 - 10 1640 92
NM_001258707.2 NP_001245636.1 9 - 10 1640 92
NM_001258708.2 NP_001245637.1 9 - 10 1640 92
NM_001258709.2 NP_001245638.1 8 - 9 1567 92
NM_001258710.2 NP_001245639.1 9 - 10 1640 92
NM_001272548.1 NP_001259477.1 8 - 9 1570 92
NM_078578.5 NP_511133.2 9 - 10 1640 92
NM_206694.4 NP_996417.1 9 - 10 1650 92
NM_206697.4 NP_996420.1 9 - 10 1640 92

View Full Product Details