Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ari-1
Entrez Gene ID
32796
Gene Name
ariadne 1
Gene Aliases
AAS65398.1, ARI1_DROME, Ari-1, CG5659, Dmel\CG5659, Dmel_CG5659, HL-V, ari, ari-1a, l(1)16Fj
Chromosome Location
Chr.X: 18130392 - 18137784 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0017418
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258807.3 NP_001245736.1 3 - 4 879 54
NM_001272742.2 NP_001259671.1 3 - 4 974 54
NM_001272743.1 NP_001259672.1 3 - 4 1090 54
NM_078675.4 NP_523399.1 3 - 4 879 54
NM_167610.4 NP_728145.1 2 - 3 858 54
NM_206777.2 NP_996500.1 3 - 4 1034 54
GenBank mRNA X98310.1 - - 828 54

View Full Product Details