Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
galectin
Entrez Gene ID
33162
Gene Name
CG11372-PA
Gene Aliases
CG11372, Dmel\CG11372, Dmel_CG11372, Dmgal
Chromosome Location
Chr.2L: 71757 - 76211 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031213
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134643.4 NP_608487.1 1 - 2 592 64
GenBank mRNA AF338142.1 - 1 - 2 376 64
AY071294.1 - 1 - 2 591 64

View Full Product Details