Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GluRIIC
- Entrez Gene ID
-
33275
- Gene Name
- Glutamate receptor IIC
- Gene Aliases
-
CG4226, CT13021, DGluR-IIC, DGluRIIC, DGluRIII, DmelGluRIIC, Dmel\CG4226, Dmel_CG4226, GLURIIC, GluR, GluR3, GluRC, GluRII-C, GluRIIC/III, GluRIII, GlutRIII, glutamate receptor III
- Chromosome Location
-
Chr.2L: 914086 - 917805 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046113
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134713.4 |
NP_608557.4 |
9 - 10 |
1982 |
62 |
View Full Product Details