Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
capt
Entrez Gene ID
45233
Gene Name
capulet
Gene Aliases
Acap, BcDNA:LD24380, CAP, CG33979, CG5061, CG5074, CP, Cap, Capt, Capulet, Dcap, Dmel\CG33979, Dmel_CG33979, acu, cap, capping protein, cyclase-associated protein, l(2)06955, punc
Chromosome Location
Chr.2L: 1151678 - 1158262 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261458
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038781.2 NP_001033870.1 1 - 2 573 70
GenBank mRNA BT021236.1 - 1 - 2 569 70

View Full Product Details