The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258911.2 | NP_001245840.1 | 4 - 5 | 918 | 66 |
NM_001258912.2 | NP_001245841.1 | 4 - 5 | 874 | 66 | |
NM_134771.4 | NP_608615.1 | 4 - 5 | 939 | 66 | |
GenBank mRNA | AY058735.1 | - | 4 - 5 | 885 | 66 |