Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pgant2
Entrez Gene ID
33556
Gene Name
Polypeptide N-Acetylgalactosaminyltransferase 2
Gene Aliases
CG3254, Dmel\CG3254, Dmel_CG3254, PGANT2, pgant2
Chromosome Location
Chr.2L: 3404460 - 3426719 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031530
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134929.4 NP_608773.2 7 - 8 1927 62
NM_205903.3 NP_995625.2 7 - 8 1963 62

View Full Product Details