Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp
Entrez Gene ID
33941
Gene Name
Thrombospondin
Gene Aliases
CG11326, CT31613, D-TSP, DTSP, Dmel\CG11326, Dmel_CG11326, SP295, TSP, tsp
Chromosome Location
Chr.2L: 6686791 - 6709086 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031850
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144330.2 NP_001137802.1 5 - 6 1042 77
NM_078771.3 NP_523495.2 4 - 5 1377 77
NM_164702.2 NP_723203.1 5 - 6 916 77
NM_164703.2 NP_723204.1 5 - 6 903 77
NM_164704.2 NP_723205.1 4 - 5 893 77
GenBank mRNA AY069229.1 - 4 - 5 875 77

View Full Product Details