Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cka
Entrez Gene ID
34096
Gene Name
Connector of kinase to AP-1
Gene Aliases
5836, CG7392, CKA, Dcka, Dmel\CG7392, Dmel_CG7392, anon-WO0172774.174, anon-WO0172774.175, anon-WO0172774.177, anon-WO0172774.179, cka, l(2)05836, l(2)s1883
Chromosome Location
Chr.2L: 8030721 - 8040826 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0044323
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259007.2 NP_001245936.1 3 - 4 1179 103
NM_001259008.2 NP_001245937.1 3 - 4 1359 103
NM_135333.3 NP_609177.1 3 - 4 1646 103
NM_164779.3 NP_723334.1 3 - 4 1179 103
NM_164780.3 NP_723335.1 3 - 4 1626 103
NM_164781.3 NP_723336.1 3 - 4 1375 103
GenBank mRNA AF364305.1 - 3 - 4 1628 103
AY061460.1 - 3 - 4 1153 103
BT120179.1 - 3 - 4 1152 103

View Full Product Details