Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jp
Entrez Gene ID
34274
Gene Name
junctophilin
Gene Aliases
CG13115, CG4405, Dmel\CG4405, Dmel_CG4405, JP
Chromosome Location
Chr.2L: 9547720 - 9566750 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032129
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201828.2 NP_001188757.1 4 - 5 1344 63
NM_001201829.2 NP_001188758.1 4 - 5 1280 63
NM_001382120.1 NP_001369124.1 5 - 6 2239 63
NM_078801.6 NP_523525.2 4 - 5 1685 63
NM_164858.2 NP_723468.1 4 - 5 1568 63
GenBank mRNA AY119554.1 - 4 - 5 1568 63

View Full Product Details