Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Marcal1
Entrez Gene ID
33709
Gene Name
CG3753-PA
Gene Aliases
CG3753, Dmel\CG3753, Dmel_CG3753, HARP, MARCAL1, Marca1, dmHARP
Chromosome Location
Chr.2L: 4955509 - 4958292 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031655
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135039.2 NP_608883.1 2 - 3 1330 143
GenBank mRNA BT011033.1 - 2 - 3 1326 143

View Full Product Details