Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp28d2
- Entrez Gene ID
-
33748
- Gene Name
- CG6081-PA
- Gene Aliases
-
28d2, CG6081, Cyp28D2, Dmel\CG6081, Dmel_CG6081
- Chromosome Location
-
Chr.2L: 5207267 - 5209345 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031688
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135067.3 |
NP_608911.1 |
5 - 6 |
1385 |
78 |
View Full Product Details