Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp28d1
Entrez Gene ID
33749
Gene Name
CG10833-PA
Gene Aliases
28d1, CG10833, CYP28d1, Dmel\CG10833, Dmel_CG10833
Chromosome Location
Chr.2L: 5210460 - 5212445 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031689
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135068.4 NP_608912.1 3 - 4 484 66
GenBank mRNA AY069176.1 - 3 - 4 465 66

View Full Product Details