Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spt20
Entrez Gene ID
39522
Gene Name
CG17689-PA
Gene Aliases
CG17689, Dmel\CG17689, Dmel_CG17689, dSpt20, lincRNA.S5070
Chromosome Location
Chr.3L: 13502707 - 13511541 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036374
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038916.3 NP_001034005.2 2 - 3 1621 71
NM_001300133.1 NP_001287062.1 2 - 3 1621 71
NM_140402.5 NP_648659.2 2 - 3 1621 71
GenBank mRNA BT016098.1 - - 1590 71

View Full Product Details