Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMS1
Entrez Gene ID
39827
Gene Name
CG4672-PA
Gene Aliases
CG4672, Dmel\CG4672, Dmel_CG4672, TMS1d
Chromosome Location
Chr.3L: 16412722 - 16415386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028399
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275018.1 NP_001261947.1 3 - 4 830 107
NM_001275019.1 NP_001261948.1 3 - 4 830 107
NM_001275020.1 NP_001261949.1 3 - 4 830 107
NM_001275021.1 NP_001261950.1 3 - 4 830 107
NM_001275022.1 NP_001261951.1 3 - 4 830 107
NM_140636.3 NP_648893.1 3 - 4 830 107
GenBank mRNA AY058257.1 - 3 - 4 797 107

View Full Product Details