Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papst2
Entrez Gene ID
39914
Gene Name
PAPS transporter 2
Gene Aliases
CG7853, Dmel\CG7853, Dmel_CG7853, PAPST2, dPAPST2
Chromosome Location
Chr.3L: 17042797 - 17044465 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036695
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140697.4 NP_648954.1 1 - 2 1120 58
GenBank mRNA AB242867.1 - 1 - 2 851 58
AY069640.1 - 1 - 2 1075 58

View Full Product Details