Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NUCB1
- Entrez Gene ID
-
39978
- Gene Name
- CG32190-PA
- Gene Aliases
-
CG32190, CG5587, Dmel\CG32190, Dmel_CG32190, NucB1, nucb1
- Chromosome Location
-
Chr.3L: 17797738 - 17802279 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052190
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140751.4 |
NP_649008.2 |
1 - 2 |
390 |
69 |
View Full Product Details