Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cni
Entrez Gene ID
34967
Gene Name
cornichon
Gene Aliases
BG:DS02740.13, CG5855, Cni, Dmel\CG5855, Dmel_CG5855, IV, anon-35Fe, spnG
Chromosome Location
Chr.2L: 16309292 - 16310726 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000339
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273566.1 NP_001260495.1 3 - 4 346 70
NM_057720.3 NP_477068.1 2 - 3 396 70
NM_165151.2 NP_723959.1 3 - 4 329 70
GenBank mRNA BT011529.1 - 3 - 4 326 70
KX531432.1 - 2 - 3 260 70
U28069.1 - 2 - 3 352 70

View Full Product Details