Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mnb
- Entrez Gene ID
-
32771
- Gene Name
- minibrain
- Gene Aliases
-
CG42273, CG7826, CG7835, DYRK1, Dm1, DmDyrk1A, Dmel\CG42273, Dmel_CG42273, Dyrk1, Dyrk1A, ME-IV, MNB, Mnb, min
- Chromosome Location
-
Chr.X: 17862697 - 17887222 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259168
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170658.3 |
NP_728106.1 |
8 - 9 |
3658 |
92 |
View Full Product Details