Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- w-cup
- Entrez Gene ID
-
34455
- Gene Name
- world cup
- Gene Aliases
-
BcDNA:AT31112, CG7363, Dmel\CG7363, Dmel_CG7363
- Chromosome Location
-
Chr.2L: 10575860 - 10577792 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032269
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135579.3 |
NP_609423.1 |
1 - 2 |
1036 |
70 |
View Full Product Details