Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lim3
Entrez Gene ID
35184
Gene Name
CG10699-PA
Gene Aliases
CG10699, CG15174, Dmel\CG10699, Dmel_CG10699, LIM3, Lim-3A, Lim3A, l(2)37Bd, l(2)Bd, lim3
Chromosome Location
Chr.2L: 19079256 - 19108427 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002023
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259156.1 NP_001246085.1 7 - 8 1764 73
NM_001259157.3 NP_001246086.1 6 - 7 1475 73
NM_001259158.3 NP_001246087.1 6 - 7 1047 73
NM_001259159.1 NP_001246088.1 7 - 8 2406 73
NM_001273630.2 NP_001260559.1 6 - 7 1475 73
NM_057258.5 NP_476606.1 6 - 7 1325 73
NM_165277.3 NP_724161.1 7 - 8 1440 73
GenBank mRNA BT029915.1 - 7 - 8 1437 73

View Full Product Details