Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhe2
Entrez Gene ID
35376
Gene Name
Na[+]/H[+] hydrogen exchanger 2
Gene Aliases
CG9255, CG9256, DNhe2, DmNHE2, Dmel\CG9256, Dmel_CG9256, Dnhe2, NHE2, dNhe2, nhe2
Chromosome Location
Chr.2L: 21104568 - 21134255 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040297
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144375.3 NP_001137847.1 17 - 18 3353 109
NM_001144377.3 NP_001137849.1 17 - 18 3377 109
NM_001144379.3 NP_001137851.1 18 - 19 3662 109
NM_001259187.2 NP_001246116.1 18 - 19 3569 109
NM_001259188.2 NP_001246117.1 18 - 19 3662 109
NM_001273731.1 NP_001260660.1 19 - 20 3207 109
NM_001273732.1 NP_001260661.1 17 - 18 3407 109
NM_136244.5 NP_610088.2 18 - 19 3569 109
NM_165355.4 NP_724311.1 18 - 19 3563 109

Gene Symbol
CG46307
Entrez Gene ID
30522396
Gene Name
CG46307-PA
Gene Aliases
Dmel\CG46307, Dmel_CG46307, Nhe2-5'utr
Chromosome Location
Chr.2L: 21104568 - 21132909 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284223
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347785.1 NP_001334745.1 17 - 18 3377 109

View Full Product Details