Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
koi
Entrez Gene ID
35594
Gene Name
klaroid
Gene Aliases
BEST:LD11166, CG18584, CG3287, CG44154, Dmel\CG44154, Dmel_CG44154, Koi, n(2)09967
Chromosome Location
Chr.2R: 6811019 - 6818199 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265003
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273811.1 NP_001260740.1 3 - 4 1210 78
NM_165491.3 NP_724502.1 4 - 5 1251 78
NM_165492.3 NP_724503.2 3 - 4 1077 78

View Full Product Details