Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tsp42Eh
- Entrez Gene ID
-
35617
- Gene Name
- Tetraspanin 42Eh
- Gene Aliases
-
CG12844, Dm.Tsp42Eh, Dmel\CG12844, Dmel_CG12844
- Chromosome Location
-
Chr.2R: 7033562 - 7034920 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033129
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078910.4 |
NP_523634.1 |
4 - 5 |
711 |
78 |
View Full Product Details