Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wun
Entrez Gene ID
35966
Gene Name
wunen
Gene Aliases
CG8804, Dmel\CG8804, Dmel_CG8804, LPP-like, Wun, Wunen, l(2)k16806
Chromosome Location
Chr.2R: 9400531 - 9413463 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016078
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259294.2 NP_001246223.1 5 - 6 1237 61
NM_057845.5 NP_477193.1 4 - 5 837 61
NM_165674.3 NP_724787.1 5 - 6 1237 61
GenBank mRNA AF145595.1 - 4 - 5 821 61
BT001729.1 - 5 - 6 1234 61

View Full Product Details