Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sn
- Entrez Gene ID
-
31717
- Gene Name
- singed
- Gene Aliases
-
CG15331, CG1536, CG32858, Dmel\CG32858, Dmel_CG32858, SN, Sn, fascin, fs(1)A1057, fs(1)K1421, fs(1)K418, fs(1)K473, fs(1)K743, fs(1)M45
- Chromosome Location
-
Chr.X: 7964024 - 7986601 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003447
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078521.3 |
NP_511076.3 |
1 - 2 |
144 |
69 |
View Full Product Details