Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sn
Entrez Gene ID
31717
Gene Name
singed
Gene Aliases
CG15331, CG1536, CG32858, Dmel\CG32858, Dmel_CG32858, SN, Sn, fascin, fs(1)A1057, fs(1)K1421, fs(1)K418, fs(1)K473, fs(1)K743, fs(1)M45
Chromosome Location
Chr.X: 7964024 - 7986601 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169226.2 NP_001162697.1 5 - 6 1805 55
NM_078521.3 NP_511076.3 5 - 6 1676 55
NM_167142.3 NP_727226.1 6 - 7 1939 55
NM_167143.3 NP_727227.1 5 - 6 2196 55

View Full Product Details