Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- btd
- Entrez Gene ID
-
31912
- Gene Name
- buttonhead
- Gene Aliases
-
BTD, Btd, CG12653, Dmel\CG12653, Dmel_CG12653
- Chromosome Location
-
Chr.X: 9694186 - 9697558 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000233
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078545.3 |
NP_511100.1 |
1 - 2 |
1225 |
59 |
View Full Product Details