Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Idgf4
Entrez Gene ID
31926
Gene Name
Imaginal disc growth factor 4
Gene Aliases
CG1780, DmIDGF4, Dmel\CG1780, Dmel_CG1780, IDGF, IDGF-4, IDGF4, IGDF4, idgf4
Chromosome Location
Chr.X: 9927973 - 9930948 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026415
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298139.1 NP_001285068.1 2 - 3 162 90
NM_078546.4 NP_511101.2 2 - 3 162 90
NM_167207.3 NP_727374.1 2 - 3 189 90
GenBank mRNA AY070943.1 - 2 - 3 193 90
BT003236.1 - - 664 90

View Full Product Details