Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hiw
Entrez Gene ID
32429
Gene Name
highwire
Gene Aliases
CG32592, CG9041, CG9049, Dmel\CG32592, Dmel_CG32592, HIW, Hiw
Chromosome Location
Chr.X: 15010137 - 15063429 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030600
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298329.1 NP_001285258.1 13 - 14 8695 79
NM_078604.4 NP_511159.3 13 - 14 8695 79

View Full Product Details