Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hiw
Entrez Gene ID
32429
Gene Name
highwire
Gene Aliases
CG32592, CG9041, CG9049, Dmel\CG32592, Dmel_CG32592, HIW, Hiw
Chromosome Location
Chr.X: 15010137 - 15063429 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030600
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298329.1 NP_001285258.1 15 - 16 9418 60
NM_078604.4 NP_511159.3 15 - 16 9418 60

View Full Product Details