Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nonA
Entrez Gene ID
32603
Gene Name
no on or off transient A
Gene Aliases
Bj6, Bj6-1, CG4211, Dmel\CG4211, Dmel_CG4211, NONA, NonA, dNONO, diss, non-A, nonA/diss, opm2, p54[nrb], x14
Chromosome Location
Chr.X: 16361569 - 16371547 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004227
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298395.1 NP_001285324.1 4 - 5 2125 131
NM_078643.3 NP_523367.2 4 - 5 2125 131
GenBank mRNA AY119651.1 - 4 - 5 2142 131

View Full Product Details